Palladium 10% on carbon, tørr for use as a catalyst Type 487
Leverandør: Thermo Scientific
Warning
A12012.04 A12012.06 A12012.09 A12012.14 A12012.18
A12012.04EA 1625 NOK
A12012.04 A12012.06 A12012.09 A12012.14 A12012.18
Palladium 10% on carbon, tørr for use as a catalyst Type 487
Palladium
CAS No.: 7440-05-3 Merck Reference: 14,6989 UN No.: 1325 MDL No.: MFCD00011167
Formel: Pd
MW: 106,420 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011167
CAS-nummer: 7440-05-3
EINECS: 231-115-6
FN: 1325
ADR: 4.1,III
Merck Indeks: 14,6989