Methylenediphosphonic acid ≥99%

Leverandør: Thermo Scientific
Danger

Synonymer: Medronic acid

A14803.03 A14803.06 A14803.14
A14803.03EA 1100 NOK
A14803.03 A14803.06 A14803.14
Methylenediphosphonic acid ≥99%
Methylenediphosphonic acid

Purity: 99+% CAS No.: 1984-15-2 Molecular Formula: CH6O6P2 Formula Weight: 176.00 Odour: Odourless Melting Point: 202-204° Beilstein Reference: 1708494 Merck Reference: 14,5792 UN No.: 3261 MDL No.: MFCD00002139

Formel: CH₆O₆P₂
MW: 176 g/mol
Smeltepunkt: 202…204 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00002139
CAS-nummer: 1984-15-2
EINECS: 217-851-0
FN: 3261
ADR: 8,III
Merck Indeks: 14,5792

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR