Adamantan 99+%
Leverandør: Thermo Scientific
|
Warning
|
146110250
146111000
146110050
146110010
ACRO146110250EA
390
NOK
ACRO146110250
ACRO146111000
ACRO146110050
ACRO146110010
Adamantan 99+%
Adamantane
Formel:
C₁₀H₁₆ MW: 136,24 g/mol Kokepunkt: 171 °C (1013 hPa) Smeltepunkt: 205…210 °C (subl.) Tetthet: 1,07 g/cm³ (20 °C) Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00074719 CAS-nummer: 281-23-2 EINECS: 206-001-4 FN: 3077 ADR: 9,III Merck Indeks: 13,00149 |
Spesifikasjon Testresultater
Appearance | White to beige Crystalline powder or crystals |
Infrared spectrum | Conforms |
GC | ≥99.0 % |
HPLC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...