TLC-plater, silikagel, 60G, glass
Leverandør: Merck
1.00384.0001 1.00390.0001
1.00384.0001EA 1850 NOK
1.00384.0001 1.00390.0001
TLC-plater, silikagel, 60G, glass
TLC-plater
Highly robust plates with gypsum as binder, fully compliant with international pharmacopoeia (Ph. Eur. and USP).
Traditionally, TLC monographs in a pharmacopoeia refer to products using silica G, containing gypsum as binder, or silica H with no foreign binder. There are about 200 monograph methods in the European Pharmacopoeia (Ph. Eur.) referring to these plates*. Fully compliant with international pharmacopoeia, these new TLC silica gel 60G plates are recommended for customers in QA/QC labs using older Ph. Eur. monograph methods, which require TLC plates with a gypsum binder and who doMore Product Information