Filling scoops for industry applications
Leverandør: BURKLE
8300-5001 8300-1002 8300-5002
BURK8300-5001EA 225 NOK
BURK8300-5001 BURK8300-1002 BURK8300-5002
Filling scoops for industry applications
Øser
PP, green. These durable scoops are ideal for use in the pharmaceutical industry, biotechnology, chemistry, industry, agriculture, fish farming, sampling, veterinary use.
  • High chemical resistance
  • Very durable
  • Anti-static
  • Autoclavable at 121 °C