Hansker, for generell bruk, Soflex
Leverandør: HONEYWELL SAFETY
2095225/10 2095225/8 2095225/9
112-1511EA 0 NOK
112-1511 112-1514 112-1515
Hansker, for generell bruk, Soflex
Hansker
Nitrile glove on interlock cotton base with ventilated back and knitted wrist. The nitrile coating ensures good chemical properties and outstanding mechanical properties (abrasion).
  • Nitrile formula gives greater elasticity and unrivalled dexterity
  • Cotton interlock lining strengthens the glove's mechanical performance while absorbing hand perspiration
  • Ventilated back improves the glove's breathability
  • Ribbed at the cuff, good wrist support is provided, thus eliminating risks of getting caught
Particularly suitable for the light and medium handling ofMore Product Information
Bestill nå
SPESIFIKASJONER
Ambidekstral/håndspesifikk Håndspesifikk
Environmentally Preferable
Engangs/flergangs gjenbrukbar
Linermateriale Bomull
Overflate ¾ coating
Type mansjett Strikket håndledd
EN norm EN 388:2016 (3111X)