GC ferrules, SilTite™ FingerTite
Leverandør: Trajan Scientific and Medical
073612 073633 073630 073631 073636 073640 073623 073622 073619 073621 073610 073618 073620 0736100 0736102 0736104 0736105 073615 073616 07361053 073632 073651
568-0309EA 5225 NOK
568-0309 568-0323 568-0321 568-0322 568-0324 568-0325 568-0318 568-0317 568-0314 568-0316 568-0302 568-0313 568-0315 568-0303 568-0305 568-0307 568-0308 568-0311 568-0312 568-0488 568-0626 568-0691
GC ferrules, SilTite™ FingerTite
Fittings og kontakter
Designed to be installed only with the force of your fingers delivering an easy, leak-free installation for capillary columns without the use of any tools. Developed with the unique, leak- and air-free properties of SilTite™ ferrules, which results in superior air tightness to reduce background noise in sensitive MS applications.
These ferrules require a unique adapter to enable finger-force connection. No permanent instrument modifications are required, so if you need to switch back to conventional connections this can be done quickly and easily.
Bestillingsinformasjon: Each starterMore Product Information