Tetrakis(dimethylamino)ethylene ≥95.0% (by GC)

Leverandør: TCI
Danger

Synonymer: TDAE

T1221-25G T1221-5G
TCIAT1221-25GEA 55675 NOK
TCIAT1221-25G TCIAT1221-5G
Tetrakis(dimethylamino)ethylene ≥95.0% (by GC)
Tetrakis(dimethylamino)ethylene
Formel: C₁₀H₂₄N₄
MW: 200,33 g/mol
Kokepunkt: 59°C
Flash punkt: 53°C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00009601
CAS-nummer: 996-70-3
EINECS: 213-638-1
FN: 2734
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a8fe4871059c33251b828c4fd9fdb3fe2003a164&vid=a8fe4871059c33251b828c4fd9fdb3fe2003a164&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Yellow to Orange clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR