VWR®, Calibration Weights, Class F2
Leverandør: VWR Collection
611-8095EA 2175 NOK
611-8095 611-8096 611-8097 611-8098 611-8099 611-8100 611-8125 611-8126 611-8127 611-8128 611-8129 611-8130 611-8154 611-8155 611-8156 611-8157 611-8158 611-8159 611-8160 611-8161 611-8162 611-8163 611-8164 611-8165 611-8166 611-8167 611-8168 611-8169 611-8170 611-8171 611-8172 611-8173 611-8174 611-8175 611-8176 611-8177 611-8178 611-8179 611-8180 611-8181 611-8182 611-8183 611-8184 611-8185 611-8186 611-8187 611-8188 611-8189 611-8190 611-8195 611-8196 611-8197 611-8198 611-8199 611-8200 611-8223 611-8224 611-8225 611-8226 611-8227
VWR®, Calibration Weights, Class F2
Veielodd Kalibreringslodd
Stainless steel Class F2 weights for users who require moderate levels of tolerance values in their calibration and day-to-day weighing.
  • Wire weights up to 500 mg in cobalt alloy
  • Identification number guarantees traceability for weights from 1 g upwards
  • Laser engraving improves legibility of the markings
  • Available with or without COFRAC certificate
Sertifiseringer: Conform to OILM R111 recommendations.
Bestillingsinformasjon: All weights from 1 mg to 20 kg are available with or without COFRAC certificate - please visit website or contact your localMore Product Information