Visual immunoassays
Leverandør: TECRA
BDEVIA48
CAMVIA96
ECOVIA48
LISVIA96
PSDVIA96
SALVIA96
SETVIA96
SIDVIA72
709-0260EA
0
NOK
709-0260
709-0261
709-0262
709-0269
709-0259
709-0253
709-0255
709-0256
Visual immunoassays
Mikrobiologiske testsystemer
Antigen-antistoff tester
Ag-AK, Immunoassay
Microtitre-format screening tests with high-affinity, specific antibodies for food and environment samples. Fast, specific bacteria and toxin testing with advanced Visual Immunoassay for food, water, environment samples, raw materials, cosmetics and pharmaceuticals. Ideal for rapid detection of pathogenic bacteria, 46 or 94 tests per plate.
- The simplicity of the VIA™ method frees up time for other work and projects
- Rapid results allow products to be released earlier, thereby saving on warehousing costs
- The kit can be used manually, semi automatically or fully automatically for large numbers of samples
- Fast results
- High sensitivity and specificity