jetPRIME® DNA and siRNA Transfection Reagents
Leverandør: SARTORIUS
101000001 101000015 101000027 101000046 201000003
101000001.EA 60375 NOK
101000001. 101000015. 101000027. 101000046. 201000003.
jetPRIME® DNA and siRNA Transfection Reagents
Transfeksjonsreagenser
The jetPRIME® DNA and/or siRNA transfection reagent is a powerful and versatile reagent designed to ensure high DNA transfection efficiency and excellent gene silencing in a variety of adherent cells.
  • High DNA transfection efficiency
  • Uses low amounts of nucleic acid
  • Superior cell viability
  • One reagent for DNA and/or siRNA transfection
  • Cost effective
jetPRIME® is ideal for co-transfection of DNA and siRNA. jetPRIME® is gentle to cells since it requires low amounts of reagent and nucleic acid during transfection. Effective and non toxic DNA and siRNA delivery is essential for reliable scientific results. Free of charge samples are available upon request. Please contact your local Avantor sales office for further details.
Since 1st of September 2021 Polyplus-Tranfection changed product part numbers. Products are remaining the same and the change is only related to the product part number. New product part numbers have symbol 'New'. For duplicated products with symbol 'telephone' please be informed that it’s the same product but with previous part number which we will only offer till the end of stock.
Sertifiseringer: Polyplus-transfection® has been an ISO 9001 accredited company since 2002. Since then this certification has been successfully renewed every three years.
Leveringsinformasjon: Supplied with an optimised sterile buffer which must be used to ensure successful transfection experiments. The concentrated buffer provided with 114-75C must be diluted 1:5 in sterile H₂O just prior to use. 1,5 ml is sufficient to perform ~375 transfections in 6-well plates.