VWR®, Calibration Weight Sets, Class F1
Leverandør: VWR Collection
611-8603EA 12075 NOK
611-8603 611-8292 611-8604 611-8605 611-8606 611-8607 611-8608 611-8610 611-8611 611-8612 611-8613 611-8614 611-8615 611-8616 611-8618 611-8619 611-8620 611-8621 611-8632 611-8633 611-8634 611-8635 611-8636 611-8637 611-8638 611-8639 611-8641 611-8642 611-8643 611-8644 611-8645 611-8646 611-8647 611-8649 611-8650 611-8651 611-8652 611-8602 611-8461 611-8462 611-8463 611-8464 611-8465 611-8466 611-8467 611-4934 611-4935 611-4940 611-4941
VWR®, Calibration Weight Sets, Class F1
Veielodd Kalibreringslodd
Weights made from anti-magnetic, polished stainless steel. Suitable for medium resolution, precision electronic balances. Weights of more than 20 g have adjustment cavities.
  • Wire weights up to 500 mg in cobalt alloy
  • Identification number guarantees traceability for weights from 1 g upwards
  • Laser engraving improves legibility of the markings
  • Available with or without COFRAC certificate
Conform to OILM R111 recommendations.
Sertifiseringer: Conform to OILM R111 recommendations.
Bestillingsinformasjon: All weight sets listed in the 'set contents'More Product Information