Post-mortem scalpel handles
Leverandør: SWANN MORTON
2650 2652 2653 2609
233-0115EA 2400 NOK
233-0115 233-0116 233-0155 SWAN2609
Post-mortem scalpel handles
Skalpellblad og skaft Skalpellskaft
A versatile and comprehensive range for morticians and pathologists for use in autopsy, histopathology and general post-mortem procedures.
  • PM40 range with traditional screw fitting
  • PM60 range with standard blade fitting
PM40B and PM60B blades are 'bull nose' blades for added safety and protection against accidental sharps injuries.
Forpakning: The PM8 and PM40 stainless steel handles and all blades are supplied individually wrapped.