Laboratory jacks
Leverandør: USBECK Laborgeräte
5260/4 5262/4 5263/4
245-0022EA 9500 NOK
245-0022 245-0023 245-0024
Laboratory jacks
Laboratoriejekker
Stainless steel plates, slip-proof rubber feet. With 10 mm hole in the top plate for attaching stand rods.
  • Height adjustment by hand or cordless screwdriver
  • Integrated safety clutch
  • Shear resistant pattern
Sertifiseringer: DIN 12897
Bestillingsinformasjon: Supplied with suitable ¼" bit for cordless screwdriver. Cordless screwdriver is included in the delivery.