VitraPOR® Micro-Filter Candle-Blanks
Leverandør: ROBU GLASFILTER
18043 18014 18000 18001 18002 18003 18004 18005 18013 18015 18020 18025 18030 18031 18032 18033 18034 18040 18041 18042 18012 18044 18010 18011 18021 18022 18023 18024
ROBU18043EA 8675 NOK
ROBU18043 ROBU18014 ROBU18000 ROBU18001 ROBU18002 ROBU18003 ROBU18004 ROBU18005 ROBU18013 ROBU18015 ROBU18020 ROBU18025 ROBU18030 ROBU18031 ROBU18032 ROBU18033 ROBU18034 ROBU18040 ROBU18041 ROBU18042 ROBU18012 ROBU18044 ROBU18010 ROBU18011 ROBU18021 ROBU18022 ROBU18023 ROBU18024
VitraPOR® Micro-Filter Candle-Blanks
Filter Filterrør
VitraPOR® micro-filter candle blanks are ideal for integration into customised filter systems. Compact and chemically resistant, they provide reliable fine filtration in small volumes.
  • VitraPOR® porous filter
  • Compact dimensions
  • Ideal for custom assemblies
  • High chemical and thermal resistance
  • Suitable for fine filtration
  • Customisable connection options
  • Reusable and durable
  • Material: Borosilicate glass 3.3
VitraPOR Micro-Filter Candle Blanks are designed for precision filtration tasks in compact or customised systems. Each blank features a VitraPOR