UV Meters, analog, J-221 & J-225
Leverandør: UVP ULTRA VIOLET PRODUCTS
97-0004-01 97-0003-01
UVPA97-0004-01EA 17200 NOK
UVPA97-0004-01 UVPA97-0003-01
UV Meters, analog, J-221 & J-225
Lysmålere
The J-Series UV meters are photovoltaic devices for taking repeatable readings of UV light sources in either the 365 nm or 254 nm UV range.
  • Assures accurate readings
  • Self-powered for maximum portability
  • 5X attenuation screen and for remote readings, a four foot (1.2 m) cord connects the sensor to the meter
Sertifiseringer: J-221 Meter complies with MIL STD 45662-A and can be found in MIL-I-6868D (NSN 6635-488-5451).
Leveringsinformasjon: Each meter comes with a sensor, attenuation screen and filter.
Bestill nå
SPESIFIKASJONER
Modell J225 J221
Environmentally Preferable
Måleområde 220 - 280 nm 300 - 400 nm
Sensor built-in-sensor built-in-sensor
Display Attenuation screen, 5X Attenuation screen, 5X
B×D×H 76×76×76 mm 76×76×76 mm
Vekt 250 g 250 g