PIPES free acid ≥98%, high purity
Leverandør: VWR Chemicals
Synonymer: Piperazine-1,4-bis(2-ethanesulphonic acid), 2,2'-(Piperazine-1,4-diyl)diethanesulphonic acid, Piperazine-N,N'-bis-2-ethanesulphonic acid, 2,2'-(Piperazine-1,4-diyl)bis(ethanesulphonic) acid
0488-100GEA 3575 NOK
0488-100G 0488-500G
PIPES free acid ≥98%, high purity
PIPES
Formel: C₈H₁₈N₂O₆S₂
MW: 302,37 g/mol
Kokepunkt: 499 °C (1013 hPa)
Smeltepunkt: 300 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00006159
CAS-nummer: 5625-37-6
EINECS: 227-057-6
Merck Indeks: 12,07633
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5192468b680c7bea85a31a131811618356a531b9&vid=ef0e3d488c68ea7f6f583fede6ebf7d2196392b7&prid=web&clid=SAPDAM
Bestill nå
Spesifikasjon Testresultater
Abs.@260nm (1%, 1 N NaOH) ≤0.05
DNase none
Heavy Metals (as Pb) <5 ppm
pKa 6.7 – 6.9
Protease none detected
Purity >98%
Residue after Ignition <0.3%
RNase none detected
Solubility (1%, 1 N NaOH) none detected