3-Kloroantranilinsyre ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Warning

Synonymer: 2-Amino-3-chlorobenzoic acid

A0786-25G A0786-5G
TCIAA0786-25GEA 2450 NOK
TCIAA0786-25G TCIAA0786-5G
3-Kloroantranilinsyre ≥98.0% (by titrimetric analysis)
3-Kloroantranilinsyre
Formel: C₇H₆ClNO₂
MW: 171,58 g/mol
Kokepunkt: 330 °C (1013 hPa)
Smeltepunkt: 194 °C
MDL nummer: MFCD00134229
CAS-nummer: 6388-47-2
EINECS: 228-996-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=388ddf089fc3554ed24feec53bd0b41c0035da4f&vid=1c0c4949e5ca0a499c6e5c21ce1f61c803aecbb1&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Yellow to Orange powder to crystal
Purity (HPLC) min. 96.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 190.0 to 194.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR