Phthaloyl dichloride ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Danger

Synonymer: Phthaloyl chloride, o-Phthaloyl dichloride

P0405-25G P0405-500G P0405-100G
TCIAP0405-25GEA 220 NOK
TCIAP0405-25G TCIAP0405-500G TCIAP0405-100G
Phthaloyl dichloride ≥98.0% (by titrimetric analysis)
Phthaloyl dichloride
Formel: C₈H₄Cl₂O₂
MW: 203,02 g/mol
Kokepunkt: 276°C
Flash punkt: 151°C
MDL nummer: MFCD00000666
CAS-nummer: 88-95-9
EINECS: 201-869-0
FN: 3265
ADR: 8,II
Merck Indeks: 14,07375
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8cd26e55e1856cde595d5076c16561ed3f9d5c58&vid=8cd26e55e1856cde595d5076c16561ed3f9d5c58&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (Argentmetric Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR