Holmium 1000 mg/L Ho in 5% salpetersyre (de Ho₂O₃) Specpure® standard til AAS
Leverandør: Thermo Fisher Scientific
88070.AE 88070.AP
88070.AEEA 760 NOK
88070.AE 88070.AP
Holmium 1000 mg/L Ho in 5% salpetersyre (de Ho₂O₃) Specpure® standard til AAS
Standarder Kationstandarder Holmium Standards
Molecular Formula: Ho₂O₃ in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011049
Sertifiseringer: Traceable to NIST