2-Pyrimidinecarbonitrile

Leverandør: Cayman Chemical

Synonymer: 2-Cyanopyrimidine

10007121-1 10007121-10 10007121-5 10007121-500
CAYM10007121-1EA 615 NOK
CAYM10007121-1 CAYM10007121-10 CAYM10007121-5 CAYM10007121-500
2-Pyrimidinecarbonitrile
2-Pyrimidinecarbonitrile
Formel: C₅H₃N₃
MW: 105,1 g/mol
MDL nummer: MFCD00160513
CAS-nummer: 14080-23-0

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR