Titan 1000 mg/L Ti in 5% salpetersyre with hydrofluoric acid (max. 1%) (fra Ti) Specpure® standard til AAS
Leverandør: Thermo Scientific
35771.AE
35771.AP
35771.AEEA
640
NOK
35771.AE
35771.AP
Titan 1000 mg/L Ti in 5% salpetersyre with hydrofluoric acid (max. 1%) (fra Ti) Specpure® standard til AAS
Standarder
Kationstandarder
Titanium Standards
Molecular Formula: Ti in 5% HNO₃/tr. HF
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011264
Sertifiseringer: Traceable to NIST