(R)-(+)-4-Benzyl-2-oxazolidinone ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: (R)-4-Benzyl-2-oxazolidinone

B1786-25G B1786-5G
TCIAB1786-25GEA 1750 NOK
TCIAB1786-25G TCIAB1786-5G
(R)-(+)-4-Benzyl-2-oxazolidinone ≥98.0% (by GC)
(R)-(+)-4-Benzyl-2-oxazolidinone
Formel: C₁₀H₁₁NO₂
MW: 177,2 g/mol
MDL nummer: MFCD00010846
CAS-nummer: 102029-44-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=192f567fd009435d447fa0c49335bdb18c180d8a&vid=192f567fd009435d447fa0c49335bdb18c180d8a&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Optical purity (LC) min. 98.0 ee%
Melting point 87.0 to 90.0 deg-C
Specific rotation [a]20/D +14.0 to +15.0 deg (C=5, MeOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR