Capper/Decapper, Skrulokk, 8-kanals
Leverandør: Thermo Fisher Scientific
Matrix®
479-1200EA 46325 NOK
479-1200 479-1067
Capper/Decapper, Skrulokk, 8-kanals
Krympetang og lokkavtaker
This capper/decapper allows ergonomic manual processing of a full rack of Matrix® or Nunc™ Cryobank or Bank-It™ tubes in less than one minute. Designed to cap tubes to the optimal torque to maintain sample integrity and prevent sample loss during storage.
  • Toggle-switch operation
  • Large ejection bar makes it easy to discard and replace used caps
Speed: 4 seconds to cap/decap 8 tubes
W×D×H: 50,8×95×251 mm
Weight: 0,44 kg
Power supply: input 100 - 240 V, 60 - 50 Hz; output 12 V, 1,0 amp
Leveringsinformasjon: Supplied with 12 V rechargeable battery and stand.