Handheld refractometers, 'PEN' series
Leverandør: Atago
3730 3741
ATAG3730EA 5025 NOK
ATAG3730 ATAG3741
Handheld refractometers, 'PEN' series
Refraktometere Refraktometere, håndholdte
Designed for continuous measurement to expand your current methods.
  • Compact design for one-handed operation
  • Automatic temperature compensation (ATC)
  • Rinse off the sample by running water on the prism
Sertifiseringer: IP 65 to rinse off the sample with running water
Leveringsinformasjon: Supplied in a storage case with battery.
Bestill nå
SPESIFIKASJONER
Modell PEN-PRO PEN-Urine S.G.
Environmentally Preferable
Vekt 70 g 70 g