4-(2-Chloroethyl)morpholine hydrochloride ≥99%
Leverandør: Thermo Scientific
Danger
Synonymer: N-(2-Chloroethyl)morpholine hydrochloride, N-(2-Chloroethyl)-Morpholinium Chloride
L05771.22 L05771.36
L05771.22EA 530 NOK
L05771.22 L05771.36
4-(2-Chloroethyl)morpholine hydrochloride ≥99%
4-(2-Chloroethyl)morpholine hydrochloride
Purity: 99% CAS No.: 3647-69-6 Molecular Formula: C6H12ClNO.HCl Formula Weight: 186.08 Melting Point: 184-186° Beilstein Reference: 3684083 UN No.: 2923 MDL No.: MFCD00012797
Formel: C₆H₁₃Cl₂NO
MW: 186,08 g/mol
Smeltepunkt: 184…186 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00012797
CAS-nummer: 3647-69-6
EINECS: 222-881-2
FN: 2923
ADR: 8,III