Safety helmets, Concept
Leverandør: CENTURION SAFETY PRODUCTS
S09EKF S09EBF S09CYF S09EGF S09CRF S09CWF S09GF S09KF S09RF S09WF S09YF S09BF
111-0865EA 0 NOK
111-0865 111-0861 111-0869 111-0863 111-0866 111-0867 111-1309 111-1310 111-1311 111-1312 111-1314 111-1307
Safety helmets, Concept
Hjelmer Sikkerhethjelm
Lightweight, comfortable and modern protective helmet made from ABS (acrylonitrile butadiene styrene) with large forehead area featuring an imprint for the company logo.
  • With extended nape area
  • Six point terylene inner cradle
  • Highly absorbent hydro-flock sweatband
  • With increased side rigidity (additional lateral deformity test)
  • Wide ventilation slots
  • Without ratchet, with full peak
Size: 510 to 630 mm
Weight: 325 g
EN 397
Sertifiseringer: EN 397