Rotating shaker, Rotator Genie™
Leverandør: SCIENTIFIC INDUSTRIES ELMIS
SI-2102 SI-2103 SI-2104 SI-2202 SI-2203 SI-2204
SCIISI-2102EA 16625 NOK
SCIISI-2102 SCIISI-2103 SCIISI-2104 SCIISI-2202 SCIISI-2203 SCIISI-2204
Rotating shaker, Rotator Genie™
Risteapparater Risteapparater, roterende/gyngende
These rotators are compact but rugged lab rotators designed to perform for a wide variety of motions which can be adjusted from gentle rolling to forceful end over end mixing. Available in two interchangeable platform configurations, providing the versatility to hold virtually any kind of containers.
  • Ideal for cold rooms and incubators (will easily fit in most incubators)
  • Standard platform with retaining bands is designed to turn 90 degrees allowing different mixing motions
  • Stainless steel magnetic platform features unique spring clip plate system
Bestill nå
SPESIFIKASJONER
Modell Rotator Genie Rotator Genie
Environmentally Preferable
Ristebevegelse Roterende Roterende
Hastighet 5 - 35 min⁻¹ 5 - 35 min⁻¹
Vinkel Roterer 360 grader, standard plattform kan vendes 90 grader Roterer 360 grader, standard plattform kan vendes 90 grader
B×D×H 175×400×400 mm 175×400×400 mm
Plattform B × D 260×260 mm 290×315 mm
Vekt 5,2 kg 5,2 kg
Maks. last 1,8 kg 1,8 kg