Vortex mixers, large capacity, Multi Vortex-Genie® and Vortex Genie® Max
Leverandør: SCIENTIFIC INDUSTRIES ELMIS
SI-M256 SI-M266 SI-M276 SI-MX202 SI-MX203 SI-MX204
444-1292EA 54450 NOK
444-1292 SCIISI-M266 SCIISI-M276 444-1299 SCIISI-MX203 SCIISI-MX204
Vortex mixers, large capacity, Multi Vortex-Genie® and Vortex Genie® Max
Risteapparater Reagensrørrister
Vortex mixers with digital speed control. These units provide accuracy, reproducibility and repeatability with complete control of speed and time. Heavy zinc die casting with specially designed feet prevents 'walking'.
  • Ideal for use with open containers due to programmable gentle ramp up to speed
  • Adjustable clamping systems make these vortex mixers easy to use and allow quick changes from one type of vessel to another
Multi Vortex-Genie® provides large capacity shaking with variable speed control from gentle orbital shaking to more vigorous vortexing.
More Product Information
Bestill nå
SPESIFIKASJONER
Modell Multi Vortex-Genie Vortex-Genie Max
Environmentally Preferable
Ristebevegelse Kretsende Kretsende
kretsløp 4 mm 3 mm
Hastighetsområde 200 - 950 min⁻¹ 500 - 1950 min⁻¹
Maks. Ø for rør (mm) 29 mm 20 mm
B×D×H 270×310×195 mm 270×310×195 mm
Vekt 11,3 kg 10,4 kg