Surface disinfectants, DAX
Leverandør: CCS CLEAN CHEMICAL
495-12 493-12 670-12
CCSC495-12EA 14825 NOK
CCSC495-12 CCSC493-12 CCSC670-12
Surface disinfectants, DAX
Rengjøringsløsninger Desinfeksjon
Alcohol based disinfectants for use on of flat smooth surfaces.
  • Ready to use formulations - no dilution necessary
  • Effective by direct contact against bacteria, fungi, and certain viruses such as HIV and hepatitis B
Dax surface disinfectant Plus is a ready-to-use solution that consists of 45% IPA, contains surfactants and can be used on visibly dirty surfaces and air-dried.
KiiltoPro MD Instru Des IPA 45+ is a medical technology product for professional use in healthcare. The product consists of 45 vol% isopropyl alcohol, surfactant (removes dirt) andMore Product Information