Spectrophotometric colorimeters, Lovibond® Model F
Leverandør: Lovibond Tintometer
180000 180270
633-0138EA 0 NOK
633-0138 TINT180270
Spectrophotometric colorimeters, Lovibond® Model F
Kolorimeter Kolorimeter med Automatisk Transmittans
The Model F is a visual colorimeter designed to optimise the use of Lovibond® glass filters in order to express colourof the sample in terms of Lovibond® Red, Yellow, Blue and Neutral. It can be used for colour measurment of products which transmit light as well as for opaque solids powders and pastes. Typical applications include including fats and fatty oils, bleached lac, liquid chemicals and pharmaceuticals.
Using the viewing tube the observer can compare the light which is transmitted through or reflected back from the sample and the light transmitted through the Lovibond glassMore Product Information
Bestill nå
SPESIFIKASJONER
Modell Model F
Lyskilde 2×12 Volt, 10 W tungsten halogen lamp
Environmentally Preferable
Lystyper Approximates to daylight
Optisk system Visual comparison of sample with colour standards in transmittance and reflectance modes
Lysvei 0,1 - 153 mm
B×D×H 330×410×230 mm
Vekt 8,3 kg