Stem castors for shelving systems, Super Erecta®
Leverandør: Metro International
5M 5MB 5MPB 5MPBGSA 5PC 5PCB 5MPGSA 5MDBGSA 5MDA 5MDBA 5MP 5MDGSA 5PCBM 5PCM 5MFA 5MFBA 5MC 5MBC 5MBESD 5MESD 5MHTN 5MHTNB 5MHTP 5MHTPB 4LD 5MPRGSA 6MPR 5MDRA 6MPB
METO5MEA 685 NOK
METO5M METO5MB METO5MPB METO5MPBGSA METO5PC METO5PCB METO5MPGSA METO5MDBGSA METO5MDA METO5MDBA METO5MP METO5MDGSA METO5PCBM METO5PCM METO5MFA METO5MFBA METO5MC METO5MBC METO5MBESD METO5MESD METO5MHTN METO5MHTNB METO5MHTP METO5MHTPB METO4LD METO5MPRGSA METO6MPR METO5MDRA METO6MPB
Stem castors for shelving systems, Super Erecta®
Hjul
Stem castors: Use with Super Erecta® posts and shelves to create a mobile shelving unit to meet special needs. Stem castors are shipped with donut bumper. 5mDA series provides extra shock absorption while the 5mP series is long-wearing and resists abrasion. 5m series is the most economical castor offered. 5mFA series are designed to dampen vibration and offer superior rollability and maneuverability.
ESD Classification: non-conductive.
Stainless steel cart-washable stem castors: Stem castors are shipped with donut bumpers.
Polymer castors: Innovative polymer stem castors offerMore Product Information