Gel permeation chromatography clean-up columns, CLNpak EV
Leverandør: Resonac
F6090001 F6090002 F6090003 F6090004 F6090005
SHOWF6090001EA 38075 NOK
SHOWF6090001 SHOWF6090002 SHOWF6090003 SHOWF6090004 SHOWF6090005
Gel permeation chromatography clean-up columns, CLNpak EV
Kromatografikolonner
The CLNpak EV are styrene divinylbenzene copolymer based columns suitable for the fractionation of residual pesticides in foods.
Bestill nå
SPESIFIKASJONER
Beskrivelse CLNpak EV-2000 CLNpak EV-G CLNpak EV-2000 AC CLNpak EV-G AC CLNpak EV-200
Environmentally Preferable
Maks. trykk 2,5 MPa (25 bar) 2,5 MPa (25 bar) 2,5 MPa (25 bar)
Temperaturgrense 40 °C 40 °C 40 °C 40 °C 40 °C