Joint clamps for standard ground joints, Cowie®
Leverandør: COWIE
030.045.1 030.012.1 030.010.1
COWI030.045.1EA 395 NOK
COWI030.045.1 COWI030.012.1 COWI030.010.1
Joint clamps for standard ground joints, Cowie®
Klemmer Klemmer til NS slip
These PTFE joint clamps with totally encapsulated steel springs can be adjusted by hand and fit standard glass joints.
  • Chemically inert and totally unaffected by laboratory chemicals
  • Temperature resistant and maintain strength right up to 280 °C
  • Can be simply adjusted by compression-extension by hand