Cetyldimethylethylammonium bromide 99%
Leverandør: Thermo Scientific
Warning
Synonymer: Ethylhexadecyldimethylammonium Bromide, Hexadecylethyldimethylammonium bromide, Ethyl(1-hexadecyl)dimethylammonium bromide, Ethylhexadecyldimethylammonium bromide, Dimethylethylhexadecylammonium bromide
211871000 211875000
ACRO211871000EA 1100 NOK
ACRO211871000 ACRO211875000
Cetyldimethylethylammonium bromide 99%
Cetyldimethylethylammonium bromide
Formel: C₂₀H₄₄BrN
MW: 378,4741 g/mol
Smeltepunkt: 190 °C (dec.)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011774
CAS-nummer: 124-03-8
EINECS: 204-672-8
ADR: 6.1,III
Bestill nå
Spesifikasjon Testresultater
Appearance White Powder
Infrared spectrum Conforms
Titration Argentometric ≥98.5 %