o-Cresolphthalein
Leverandør: Thermo Scientific
Synonymer: 3,3-Bis(4-hydroxy-3-methylphenyl)isobenzofuran-1(3H)-one
A12899.09 A12899.18 A12899.99
A12899.09EA 575 NOK
A12899.09 A12899.18 A12899.99
o-Cresolphthalein
o-Cresolphthalein
CAS No.: 596-27-0 Molecular Formula: C22H18O4 Formula Weight: 346.39 Melting Point: 223-225° Beilstein Reference: 310554 Merck Reference: 14,2576 MDL No.: MFCD00005912
Formel: C₂₂H₁₈O₄
MW: 346,39 g/mol
Smeltepunkt: 223…225 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00005912
CAS-nummer: 596-27-0
EINECS: 209-881-8
Merck Indeks: 14,2576