Ion exclusion chromatography columns, SUGAR SH
Leverandør: Resonac
F6378100 F6700080 F6378101
554-2221EA 26475 NOK
554-2221 554-2229 554-2222
Ion exclusion chromatography columns, SUGAR SH
Kromatografikolonner
Columns for simultaneous analysis of saccharides and organic acids.
  • Separates saccharides in size exclusion mode and organic acids in ion exclusion mode
  • Suitable for the analysis of uronic and aldonic acids
Bestill nå
SPESIFIKASJONER
Beskrivelse SUGAR SH1011 SUGAR SH-G SUGAR SH1821
Environmentally Preferable
Maks. trykk 5,0 MPa (50 bar) 5,0 MPa (50 bar)
Temperaturgrense 50 - 60 (≤95 °C) 50 - 60 (≤95 °C) 50 - 60 (≤95 °C)
USP kode L17, L22 L17, L22 L17, L22