Size exclusion and ligand exchange chromatography columns, SUGAR SC
Leverandør: Resonac
F6700090 F6379300 F6378102
554-2231EA 8025 NOK
554-2231 554-2226 554-2223
Size exclusion and ligand exchange chromatography columns, SUGAR SC
Kromatografikolonner
These columns separate mono and di-saccharides by combination of ligand exchange and size exclusion mode.
  • Counter ion: Ca²⁺
  • Solvents, other than water, are not required for the analysis of neutral sugars
  • Base material is styrene divinylbenzene copolymer
EP SC1011-7F columns are ideal for the analysis of sugar alcohols and saccharides in conformity with EP.
Bestill nå
SPESIFIKASJONER
Beskrivelse SUGAR SC-G 6B EP SC1011-7F SUGAR SC1011
Environmentally Preferable
Maks. trykk 2,5 MPa (25 bar) 4,0 MPa (40 bar)
pH område 3 - 7 3 - 7 3 - 7
Temperaturgrense 70 - 85 (≤95 °C) 70 - 85 (≤95 °C) 70 - 85 (≤95 °C)
USP kode L19, L22 L19, L22 L19, L22