HPLC columns, Accucore™ C8
Leverandør: Thermo Fisher Scientific
17226-012105 17226-014005 17226-013005 17226-032130 17226-033030 17226-052130 17226-102130 17226-103030 17226-104630 17226-152130 17226-153030 17226-154630
554-2565EA 6650 NOK
554-2565 554-2567 554-2566 554-2568 554-2569 554-1989 554-1991 554-2572 554-1992 554-1993 554-2573 554-1994
HPLC columns, Accucore™ C8
Kromatografikolonner
These columns offer lower hydrophobic retention than columns packed with longer alkyl length material, such as C18. Based on Core Enhanced Technology, they provide fast, high resolution separations without the elevated back pressures required by sub-2 µm particles. Improve separation speed and resolution while maintaining low backpressures using Thermo Scientific™ Accucore C8 columns.
  • Low levels of secondary interactions
  • Excellent bonded phase coverage
Rugged 2,6 µm solid core particles ensure high efficiencies and enable compatibility with both HPLCMore Product Information
Bestill nå
SPESIFIKASJONER
Beskrivelse 10038-840 10038-838 10038-836 10038-834 10038-830 10038-828 10038-822 10038-820 10038-818 10038-816 10038-814 10038-812
Environmentally Preferable
Karbonbelastning 5% 5% 5% 5% 5% 5% 5% 5% 5% 5% 5% 5%
pH område 2 - 9 2 - 9 2 - 9 2 - 9 2 - 9 2 - 9 2 - 9 2 - 9 2 - 9 2 - 9 2 - 9 2 - 9
Overflate areal 130 m²/g 130 m²/g 130 m²/g 130 m²/g 130 m²/g 130 m²/g 130 m²/g 130 m²/g 130 m²/g 130 m²/g 130 m²/g 130 m²/g