VWR®, Specimen Containers, Conical, with Screw Caps
Leverandør: VWR Collection
216-0855EA 1675 NOK
216-0855 216-0856 216-0857 216-0858 216-0859 216-0860 216-0861 216-0862 216-0863 216-0864 216-0865 216-0868 216-0869 216-1185 216-3949
VWR®, Specimen Containers, Conical, with Screw Caps
Prøvecontainere Beholdere, generelt bruk
PP.
  • Complete range available with different cap colours
  • Sterile as well as non sterile, individually packed or bulk
  • Frosted labelling area
General specimen containers. 120 ml capacity.