3-Amino-o-cresol ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

Synonymer: 3-Amino-2-methylphenol, 1-Amino-3-hydroxy-2-methylbenzene, 3-Hydroxy-2-methylaniline, 2-Amino-6-hydroxytoluene

A1824-25G A1824-5G
TCIAA1824-25GEA 5450 NOK
TCIAA1824-25G TCIAA1824-5G
3-Amino-o-cresol ≥98.0% (by GC, titration analysis)
3-Amino-o-cresol
Formel: C₇H₉NO
MW: 123,15 g/mol
MDL nummer: MFCD00007787
CAS-nummer: 53222-92-7
EINECS: 258-438-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e450f27e370c86f860af9350e7c4df6bd7e6c13d&vid=e450f27e370c86f860af9350e7c4df6bd7e6c13d&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 130.0 to 135.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR