Maleamic acid ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
|
Warning
|
Synonymer:
Maleic acid monoamide, Maleamic acid, (Z)-4-Amino-4-oxobut-2-enoic acid
M0003-25G
M0003-500G
TCIAM0003-25GEA
395
NOK
TCIAM0003-25G
TCIAM0003-500G
Maleamic acid ≥98.0% (by HPLC, titration analysis)
Maleamic acid
Formel:
C₄H₅NO₃ MW: 115.0874 g/mol |
MDL nummer:
MFCD00082354 CAS-nummer: 557-24-4 EINECS: 209-163-4 Merck Indeks: 14,05701 |
|
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Solubility in hot Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...