4-Methylumbelliferyl phosphate disodium salt (4-MUP-Na2)
Leverandør: Biotium
Synonymer: Disodium 4-methylumbelliferyl phosphate, 4-MUP
10010-1 10010
10010-1EA 15175 NOK
10010-1 10010.
4-Methylumbelliferyl phosphate disodium salt (4-MUP-Na2)
4-Methylumbelliferyl phosphate disodium salt (4-MUP-Na2)
Formel: C₁₀H₇Na₂O₆P
MW: 300,11 g/mol
MDL nummer: MFCD00037614
CAS-nummer: 22919-26-2
EINECS: 245-325-0