Octyltrimethylammonium bromide ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Warning

Synonymer: Trimethyl-1-octylammonium bromide

O0163-25G O0163-500G
TCIAO0163-25GEA 715 NOK
TCIAO0163-25G TCIAO0163-500G
Octyltrimethylammonium bromide ≥98.0% (by titrimetric analysis)
Octyltrimethylammonium bromide
Formel: C₁₁H₂₆BrN
MW: 252,24 g/mol
Smeltepunkt: 217°C
MDL nummer: MFCD00051873
CAS-nummer: 2083-68-3

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to lump
Purity (Nonaqueous Titration) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR