2',7'-Dichlorofluorescein disodium salt ≥98.0% (by titrimetric analysis)

Leverandør: TCI

Synonymer: Sodium 2',7'-dichloro-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-3',6'-bis(olate), 2',7'-Dichlorofluorescein sodium salt

D0424-1G D0424-25G
TCIAD0424-1GEA 380 NOK
TCIAD0424-1G TCIAD0424-25G
2',7'-Dichlorofluorescein disodium salt ≥98.0% (by titrimetric analysis)
2',7'-Dichlorofluorescein disodium salt
Formel: C₂₀H₈Cl₂Na₂O₅
MW: 445,160 g/mol
MDL nummer: MFCD00070511
CAS-nummer: 80471-69-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7f9fb5dc79973252bff06905bc77bc70f7fbdd2d&vid=7f9fb5dc79973252bff06905bc77bc70f7fbdd2d&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Purple to Dark purple to Brown powder to crystal
Purity (Nonaqueous Titration) min. 98.0% (calcd.on anh.substance)
Solubility in Water almost transparency
Water max. 10.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR