4-Bromo-2-furaldehyde ≥97.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 4-Bromofurfural, 4-Bromofuran-2-carbaldehyde

B1816-1G B1816-5G
TCIAB1816-1GEA 2350 NOK
TCIAB1816-1G TCIAB1816-5G
4-Bromo-2-furaldehyde ≥97.0% (by GC)
4-Bromo-2-furaldehyde
Formel: C₅H₃BrO₂
MW: 174,98 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00671504
CAS-nummer: 21921-76-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=14b3a56408572238ed647e26d804860402751730&vid=14b3a56408572238ed647e26d804860402751730&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 97.0 %
Melting point 53.0 to 57.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR