(±)-2-Bromovaleric acid ≥98.0% (by GC, titration analysis)
Leverandør: TCI
|
Danger
|
Synonymer:
(±)-2-Bromopentanoic acid, 2-Bromovalericacid
B0667-25G
B0667-500G
TCIAB0667-25GEA
395
NOK
TCIAB0667-25G
TCIAB0667-500G
(±)-2-Bromovaleric acid ≥98.0% (by GC, titration analysis)
(±)-2-Bromovaleric acid
Formel:
H₃C(CH₂)₂BrCHCOOH MW: 181,03 g/mol Kokepunkt: 136°C Smeltepunkt: 38…39 °C Tetthet: 1,475 g/cm³ (20 °C) |
MDL nummer:
MFCD00004217 CAS-nummer: 584-93-0 EINECS: 209-546-6 FN: 3265 ADR: 8,III |
Spesifikasjon Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...