4-Bromoresorcinol ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Warning
Synonymer: 4-Bromobenzene-1,3-diol
B0654-25G B0654-5G
TCIAB0654-25GEA 5525 NOK
TCIAB0654-25G TCIAB0654-5G
4-Bromoresorcinol ≥98.0% (by GC, titration analysis)
4-Bromoresorcinol
Formel: C₆H₅BrO₂
MW: 189,01 g/mol
Kokepunkt: 150°C
Smeltepunkt: 98 °C
MDL nummer: MFCD00002272
CAS-nummer: 6626-15-9
EINECS: 229-586-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b72eba01370036b9e0d5523f6098ac20de542f27&vid=b72eba01370036b9e0d5523f6098ac20de542f27&prid=web&clid=SAPDAM
Bestill nå
Spesifikasjon Testresultater
Appearance White to Light yellow to Dark green powder to crystal
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Melting point 100.0 to 104.0 deg-C