5-Nitro-2-furaldehyde ≥98.0% (by GC)

Leverandør: TCI
Danger

Synonymer: 5-Nitrofurfural

N0387-25G N0387-5G
TCIAN0387-25GEA 1100 NOK
TCIAN0387-25G TCIAN0387-5G
5-Nitro-2-furaldehyde ≥98.0% (by GC)
5-Nitro-2-furaldehyde
Formel: C₅H₃NO₄
MW: 141,08 g/mol
Kokepunkt: 272 °C (1013 hPa)
Smeltepunkt: 35 °C
Tetthet: 1,34 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003230
CAS-nummer: 698-63-5
EINECS: 211-816-3
FN: 1325
ADR: 4.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6b0f2d5cd2885e51718c584bf4058d597791503d&vid=6b0f2d5cd2885e51718c584bf4058d597791503d&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Brown powder to lump to clear liquid
Purity (GC) min. 98.0 %
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR