Metyltriklorsilan ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Danger
Synonymer: Trichloro(methyl)silane
M0450-25G M0450-500G M0450-100G
TCIAM0450-25GEA 220 NOK
TCIAM0450-25G TCIAM0450-500G TCIAM0450-100G
Metyltriklorsilan ≥98.0% (by GC, titration analysis)
Metyltriklorsilan
Formel: CH₃Cl₃Si
MW: 149,48 g/mol
Kokepunkt: 66°C
Smeltepunkt: –90°C
Tetthet: 1,27 g/cm³ (20 °C)
Flash punkt: –8°C
MDL nummer: MFCD00000481
CAS-nummer: 75-79-6
EINECS: 200-902-6
FN: 1250
ADR: 3,II
REACH: 01-2119475226-36
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7362417faf61568aca62df7b3ac9836b15fa28f3&vid=7362417faf61568aca62df7b3ac9836b15fa28f3&prid=web&clid=SAPDAM
Bestill nå
Spesifikasjon Testresultater
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %